Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Phosphatase 1 beta Rabbit anti-Human, Mouse, Rat, Clone: 5O9E7, Novus Biologicals™
SDP

Catalog No. p-7247856 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167870 20 μg
NB167869 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167870 Supplier Novus Biologicals Supplier No. NBP31638920UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Protein Phosphatase 1 beta Monoclonal antibody specifically detects Protein Phosphatase 1 beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Protein Phosphatase 1 beta
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5O9E7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 3.1.3.16, EC 3.1.3.53, MGC3672, PP1beta, PP-1Bprotein phosphatase 1, catalytic subunit, delta isoform, PPP1CD, protein phosphatase 1, catalytic subunit, beta isoform, protein phosphatase 1, catalytic subunit, beta isozyme, protein phosphatase 1-beta, protein phosphatase 1-delta, serine/threonine protein phosphatase PP1-beta catalytic subunit, serine/threonine-protein phosphatase PP1-beta catalytic subunit
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 228-327 of human Protein Phosphatase 1 beta (P62140).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5500
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.