Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein phosphatase 1F Antibody (6D11), DyLight 550, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242648R

Catalog No. NBP242648R


Only null left
Add to Cart

Description

Description

Protein phosphatase 1F Monoclonal specifically detects Protein phosphatase 1F in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Protein phosphatase 1F
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
PPM1F
Recombinant human PPM1F (1-454aa) purified from E. coli
100 μL
Apoptosis, Protein Phosphatase
9647
Store at 4C in the dark.
IgG1 κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
6D11
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Ca(2+)/calmodulin-dependent protein kinase phosphatase, CaMKPaseCaM-kinase phosphatase, EC 3.1.3.16, FEM-2, hFEM-2, KIAA0015POPX2CAMKP, Partner of PIX 2, PP2C phosphatase, Protein fem-2 homolog, protein phosphatase 1F, protein phosphatase 1F (PP2C domain containing), protein phosphatase, Mg2+/Mn2+ dependent, 1F
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.