Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein Phosphatase inhibitor 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Protein Phosphatase inhibitor 1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Protein Phosphatase inhibitor 1 Polyclonal specifically detects Protein Phosphatase inhibitor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Protein Phosphatase inhibitor 1 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
I1, I-1, IPP-1, IPP1inhibitor-1, protein phosphatase 1 regulatory subunit 1A, protein phosphatase 1, regulatory (inhibitor) subunit 1A, Protein phosphatase inhibitor 1, protein phosphatase inhibitor-1 | |
PPP1R1A | |
IgG | |
Affinity Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q13522 | |
5502 | |
This antibody was developed against a recombinant protein corresponding to amino acids: AESTETQESRPPGIPDTEVESRLGTSGTAKKTAECIPKTHERGSKEPSTKEPSTHIPPLDSKGANSV | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title