Learn More
Thermo Scientific™ Protein Stabilizing Cocktail PROMO
Description
This proprietary formulation of low-molecular weight, naturally occurring molecules helps protect proteins from environmental stresses that can otherwise lead to enzyme inactivation, aggregation and freeze-thaw damage. The Protein Stabilizing Cocktail is provided as an easily pipettable, buffered 4X concentrate. Solutions of enzymes and other proteins to which the cocktail has been added may be refrigerated or frozen for storage without losing their activity or function. Although the degree of stabilization is protein-specific, the cocktail significantly stabilizes most proteins compared with conventional buffer alone. The Protein Stabilizing Cocktail is nontoxic and does not destabilize biomolecules; however, all cocktail components can be removed by dialysis or desalting before use in downstream assays, if desired.
Highlights:
- Stabilizes most enzymes and other proteins significantly better than ordinary buffers
- Protect proteins from environmental stresses
- Does not destabilize biomolecules in downstream assays
- Components are low molecular weight and fully dialyzable
- Low-viscosity reagent is easier to pipette than 50% glycerol
- Protein classes tested include kinases, phosphatases, peroxidases, restriction enzymes, luciferases, cytokines and antibodies
Specifications
Specifications
| Product Type | Protein Stabilizing Cocktail |
| Content And Storage | Upon receipt store at 4°C |
| Form | Liquid |
| For Use With (Application) | ELISA, Enzyme Assay, Functional Studies |
| Reagent Type | Stabilizing Reagent |
| Quantity | 10 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.