Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Protein-Total Reagents

Catalog No. 23666403 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1000 mL
120 mL
5 x 300 R2mL
500 mL
8 x 50mL (1.7 oz.)
Formulation:
R1 and R2 contains Sodium Hydroxide 750mM, Copper Sulfate 6mM, Sodium Potassium Tartrate 35mM, Potassium Iodide 6mM, non-reactive ingredients
Sodium Hydroxide 600mM, Copper Sulfate 12mM, Sodium Potassium Tartrate 32mM, Potassium Iodide 30mM, non-reactive ingredients
pH:
13.15
13.2
Type:
Total Protein Reagent Kit
Total Protein Reagent Set
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation pH Type
23-666-403 8 x 50mL (1.7 oz.) R1 and R2 contains Sodium Hydroxide 750mM, Copper Sulfate 6mM, Sodium Potassium Tartrate 35mM, Potassium Iodide 6mM, non-reactive ingredients 13.2 Total Protein Reagent Kit
23-666-391 500 mL Sodium Hydroxide 600mM, Copper Sulfate 12mM, Sodium Potassium Tartrate 32mM, Potassium Iodide 30mM, non-reactive ingredients 13.15 Total Protein Reagent Set
23-666-389 120 mL Sodium Hydroxide 600mM, Copper Sulfate 12mM, Sodium Potassium Tartrate 32mM, Potassium Iodide 30mM, non-reactive ingredients 13.15 Total Protein Reagent Set
23-666-390 1000 mL Sodium Hydroxide 600mM, Copper Sulfate 12mM, Sodium Potassium Tartrate 32mM, Potassium Iodide 30mM, non-reactive ingredients 13.15 Total Protein Reagent Set
23-666-401 5 x 300 R2mL R1 and R2 contains Sodium Hydroxide 750mM, Copper Sulfate 6mM, Sodium Potassium Tartrate 35mM, Potassium Iodide 6mM, non-reactive ingredients 13.2 Total Protein Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 23-666-403 Supplier Pointe Scientific Supplier No. HT728600

Includes

Sodium hydroxide 600mM; Copper sulfate 12mM; Sodium potassium tartrate 32mM; Potassium iodide 30mM; Non-reactive ingredients

Quantify total protein in serum

  • Method: Biuret, Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 540nm
  • Linearity: 1.0 to 15.0g/dL
  • Expected Values: 6.2 to 8.5g/dL

For the quantitative determination of total protein concentration in serum

Specifications

Certifications/Compliance CE Marked
Type Total Protein Reagent Kit
Formulation R1 and R2 contains Sodium Hydroxide 750mM, Copper Sulfate 6mM, Sodium Potassium Tartrate 35mM, Potassium Iodide 6mM, non-reactive ingredients
For Use With (Application) Quantitative determination of total Protein concentration in Serum, in-vitro diagnostic use only
Model 704, 902
pH 13.2
Quantity 8 x 50mL (1.7 oz.)
Form Liquid

For in vitro diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.