Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein-tyrosine phosphatase eta (CD148) Syrian Hamster anti-Mouse, Unconjugated, Clone: 8A-1, MilliporeSigma™

Catalog No. MABF207025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2070-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2070-25 Supplier MilliporeSigma Supplier No. MABF207025UG
Add to Cart
Add to Cart

Syrian Hamster Monoclonal Antibody

Anti-Protein-tyrosine phosphatase eta (CD148), clone 8A-1, Cat. No. MABF2070, is a Syrian hamster monoclonal antibody that detects Receptor-type tyrosine-protein phosphatase eta (CD148) and is tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein-tyrosine phosphatase eta (CD148)
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8A-1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified Syrian hamster monoclonal antibody IgG2-IgG3 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Receptor-type tyrosine-protein phosphatase eta;EC: 3.1.3.48;Protein-tyrosine phosphatase eta;R-PTP-eta;HPTP beta-like tyrosine phosphatase;Protein-tyrosine phosphatase receptor type J;R-PTP-J;Susceptibility to colon cancer 1;CD148
Gene Symbols Ptprj;Byp;Scc1
Host Species Syrian Hamster
Immunogen 293T cells expressing murine CD148.
Molecular Weight of Antigen 136.77 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone 8A-1 immunostains Protein-tyrosine phosphatase eta in  murine spleen tissue secctions.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.