Learn More
Invitrogen™ Protein Tyrosine Phosphatase Polyclonal Antibody

Description
Protein Tyrosine Phosphatase Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | Protein Tyrosine Phosphatase |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.12 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Host Species | Rabbit |
| Immunogen | Recombinant Zebrafish Protein tyrosine phosphatase, receptor type, s, a protein (269-424AA). |
| Purification Method | Protein G |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.