Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Tyrosine phosphatase Receptor F (PTPRF) Monoclonal Antibody (S165-38), QED Bioscience™
SDP

Catalog No. 502278255
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8255 Supplier Leinco Technologies Inc Supplier No. 56579100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Protein Tyrosine phosphatase Receptor F (PTPRF) Monoclonal antibody specifically detects Protein Tyrosine phosphatase Receptor F (PTPRF) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen Protein Tyrosine phosphatase Receptor F (PTPRF)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone S165-38
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Gene PTPRF
Gene Accession No. A2A8L5, P10586, Q64604
Gene Alias AA591035; BNAH2; FLJ43335; FLJ45062; FLJ45567; LAR; LAR protein; LARFN5C; LARS; LCA homolog; LCA-homolog; leukocyte antigen-related (LAR) PTP receptor; leukocyte antigen-related tyrosine phosphatase; leukocyte common antigen related; protein tyrosine phosphatase receptor-type F; protein tyrosine phosphatase, receptor type F; protein tyrosine phosphatase, receptor type, F; protein tyrosine phosphatase, receptor type, F polypeptide; Ptprf; PTPRF protein; receptor-linked protein-tyrosine phosphatase LAR; receptor-type tyrosine-protein phosphatase F; RPTP-LAR
Host Species Mouse
Immunogen Fusion protein corresponding to aa 1315-1607 (cytoplasmic C-terminus) of human PTPRF. This sequence is 97
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19268, 360406, 5792
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.