Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein Z Antibody (OTI4D5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273673AF350
Description
Protein Z Monoclonal specifically detects Protein Z in Human samples. It is validated for Immunohistochemistry.Specifications
Protein Z | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
PROZ | |
Recombinant protein fragment corresponding to amino acids 115-400 of human Protein Z (NP_003882) produced in E.coli. | |
0.1 mL | |
8858.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI4D5 | |
Immunohistochemistry | |
protein Z, vitamin K-dependent plasma glycoprotein, PZ, vitamin K-dependent protein Z | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction