Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protein Z Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562529 Supplier Invitrogen™ Supplier No. PA562529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPEKDFAEHL LIPRTRGLLS GWARNGTDLG NSLTTRPVTL VEGEECGQVL NVTVTTRTYC ERSSVAAMHW MDGSVVTREH RGSWFLTGVL GSQPVGGQAH MVLVTKVSRY S Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 62%.

PROZ protein Z, vitamin K-dependent plasma glycoprotein. It is 62 kDa large and 396 amino acids long. It has four domains: a gla-rich region, two EGF-like domains and a trypsin-like domain. It lacks the serine residue that would make it catalytically active as a serine protease. It is a member of the coagulation cascade, the group of blood proteins that leads to the formation of blood clots. It is vitamin K-dependent, and its functionality is therefore impaired in warfarin therapy. It is a glycoprotein. Although it is not enzymatically active, it is structurally related to several serine proteases of the coagulation cascade: factors VII, IX, X and protein C. The carboxyglutamate residues (which require vitamin K) bind protein Z to phospholipid surfaces. The main role of protein Z appears to be the degradation of factor Xa. This is done by protein Z-related protease inhibitor (ZPI), but the reaction is accelerated 1000-fold by the presence of protein Z. Oddly, ZPI also degrades factor XI, but this reaction does not require the presence of protein Z. In some studies, deficiency states have been associated with a propensity to thrombosis. Others, however, link it to bleeding tendency; there is no clear explanation for this, as it acts physiologically as an inhibitor, and deficiency would logically have led to a predisposition for thrombosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Z
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PROZ
Gene Accession No. P22891
Gene Alias protein Z, vitamin K dependent plasma glycoprotein; protein Z, vitamin K-dependent plasma glycoprotein; PROZ; PZ; vitamin K-dependent protein Z; vitamin K-dependent protein Z precursor variant 1
Gene Symbols PROZ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Protein Z. Recombinant protein control fragment (Product #RP-103897).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8858
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.