Learn More
Invitrogen™ Protein Z Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120422
Description
Positive test controls include: HepG2, 293T, Mouse kidney, Rat liver, Rat kidney. The target is usually found in the following locations: Secreted. Immunogen sequence: SLLHRNITVK TYFNRTSQDP LMIKITHVHV HMRYDADAGE NDLSLLELEW PIQCPGAGLP VCTPEKDFAE HLLIPRTRGL LSGWARNGTD LGNSLTTRPV TLVEGEECGQ VLNVTVTTRT Y.
PROZ protein Z, vitamin K-dependent plasma glycoprotein. It is 62 kDa large and 396 amino acids long. It has four domains: a gla-rich region, two EGF-like domains and a trypsin-like domain. It lacks the serine residue that would make it catalytically active as a serine protease. It is a member of the coagulation cascade, the group of blood proteins that leads to the formation of blood clots. It is vitamin K-dependent, and its functionality is therefore impaired in warfarin therapy. It is a glycoprotein. Although it is not enzymatically active, it is structurally related to several serine proteases of the coagulation cascade: factors VII, IX, X and protein C. The carboxyglutamate residues (which require vitamin K) bind protein Z to phospholipid surfaces. The main role of protein Z appears to be the degradation of factor Xa. This is done by protein Z-related protease inhibitor (ZPI), but the reaction is accelerated 1000-fold by the presence of protein Z. Oddly, ZPI also degrades factor XI, but this reaction does not require the presence of protein Z. In some studies, deficiency states have been associated with a propensity to thrombosis. Others, however, link it to bleeding tendency; there is no clear explanation for this, as it acts physiologically as an inhibitor, and deficiency would logically have led to a predisposition for thrombosis.
Specifications
| Protein Z | |
| Polyclonal | |
| Unconjugated | |
| PROZ | |
| protein Z, vitamin K dependent plasma glycoprotein; protein Z, vitamin K-dependent plasma glycoprotein; PROZ; PZ; vitamin K-dependent protein Z; vitamin K-dependent protein Z precursor variant 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 306608, 66901, 8858 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.48 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P22891, Q9CQW3 | |
| PROZ | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-340 of human PROZ (NP_003882.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.