Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteinase 3/Myeloblastin/PRTN3 Antibody (3B4), Novus Biologicals™
SDP

Catalog No. NBP151573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151573 Supplier Novus Biologicals Supplier No. NBP151573
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Proteinase 3/Myeloblastin/PRTN3 Monoclonal specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Proteinase 3/Myeloblastin/PRTN3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3B4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000
Gene Alias ACPA, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen
Gene Symbols PRTN3
Host Species Mouse
Immunogen Purified recombinant fragment of human Proteinase 3 expressed in E. Coli.
Molecular Weight of Antigen 28 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 5657
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.