Learn More
Proteinase 3/Myeloblastin/PRTN3 Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier:  Novus Biologicals NBP125966
 View more versions of this product
Catalog No. NBP125966
Only null left
Add to Cart
Description
Description
Proteinase 3/Myeloblastin/PRTN3 Polyclonal specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Proteinase 3/Myeloblastin/PRTN3
Polyclonal
Unconjugated
Lyophilized from whole antisera with No Preservative
ACPA, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen
Rabbit
28 kDa
0.1 mL
Primary
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
LYOPH
Western Blot 1:200-1:1000, Flow Cytometry 1:200-1:1000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen 1:200-1:1000
P24158
PRTN3
A synthetic peptide from the c-terminal region of human Proteinase 3/Myeloblastin/PRTN3 conjugated to blue carrier protein was used as the antigen.
Unpurified
RUO
5657
Human
Antisera
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only