Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteinase 3/Myeloblastin/PRTN3 Antibody (SR1444), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322274100UL
Description
Proteinase 3/Myeloblastin/PRTN3 Monoclonal antibody specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Proteinase 3/Myeloblastin/PRTN3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1444 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
ACPA, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen | |
A synthesized peptide derived from human Proteinase 3/Myeloblastin/PRTN3 (Uniprot #: P24158) | |
100 μg | |
Cell Biology, Cell Cycle and Replication, Cellular Markers | |
5657 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction