Learn More
Description
Proteinase K is used in digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research. Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA, as well as in removal of endotoxins bound to cationic proteins such as lysozyme and ribonuclease A and in determination of enzyme localization on membranes.
- Grade: Molecular biology reagent
- Specific Activity: >30 mAnson u/mg
- Presentation: White powder
- Isoelectric Point: 8.9 (Lit.)
- Extinction Coefficient: E1%= 14.2 (280 nm,10 mM NaCl and 5 mM CaCl2, pH 8.0) (Lit.)
- pH: 7.5 -12.00 (denatured hemoglobin as substrate) (Lit.)
- Soluble in water
One Anson unit is the amount of enzyme which will liberate 1 °mole of folin-positive amino acids per minnute at pH 7.5 and 35°C using hemoglobin as substrat
Isolation of DNA and RNA
Specifications
Specifications
| Color | White |
| Grade | Molecular Biology |
| pH | 7.5 to 12 |
| For Use With (Application) | Useful for the proteolytic inactivation of Nucleases during the isolation of DNA and RNA |
| Content And Storage | 4°C |
| Source | Tritirachium album |
| Quantity | 5 mg |
| Product Type | Proteinase K |
| Molecular Weight (g/mol) | 30000 |
| Inhibitors | Diisopropylfluorophosphate or Phenylmethane Sulfonyl Fluoride, AEBSF and Trypsin |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.