Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Proteinase K (PK) Solution

Catalog No. PRMC5008 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
4 mL
16 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRMC5008 16 mL
PRMC5005 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRMC5008 Supplier Promega Supplier No. MC5008
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Proteinase K, produced by the fungus Tritirachium album Limber, is a serine protease that exhibits broad cleavage activity. Proteinase K (PK) Solution is supplied at a concentration of 20mg/ml in 10mM Tris-HCl (pH 7.5), 1mM calcium chloride, 50% glycerol.

  • Stable at Room Temperature and Easy to Use
  • Active over pH range 4.3-12.0 in 0.5% SDS or 1% Triton(R) X-100
  • Retains >80% activity at temperatures up to 60°C
  • No resuspension or thawing before use
  • Cat.# MC5005, MC5008 supplied at a concentration of 20mg/ml

Specifications

Concentration 20 mg/mL
pH 4.3 to 12
Quantity 16 mL
Product Type Proteinase K (PK) Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.