Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Proteinase K Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120726 Supplier Invitrogen™ Supplier No. PA5120726
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Proteinase K.

Proteinase K (E.C.3.4.21.64) is an extracellular serine endoproteinase produced by Tritirachinm album Limber. Proteinase K is active in a wide range of temperatures and buffers with optimal activity between 20 and 6°C and a pH between 7.5 and 12.0. Activity is stimulated when up to 2% SDS or up to 4 M urea are included in the reaction. Calcium is important for thermostability of Proteinase K but it is not required for catalysis, therefore Proteinase K is also active in buffers containing chelating agents such as EDTA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proteinase K
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Alias EC 3.4.21.14; Protease K
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.