Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteinase K Rabbit anti-Fungi, Polyclonal, Nordic
SDP

Catalog No. 502101547
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1547 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1547 Supplier Vector Laboratories Supplier No. NE144/7S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Prior to use, an aliquot is thawed slowly at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4 C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the product

Proteinase K Polyclonal antibody specifically detects Proteinase K in Fungi samples. It is validated for Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunofluorescence

Specifications

Antigen Proteinase K
Applications Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 10 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH
Gene Alias EC 3.4.21.14; Protease K
Host Species Rabbit
Immunogen Tritirachium album Proteinase K
Purification Method Protein G
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.