Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Proteinase K Solution, Molecular Biology Grade, 20 mg/mL

Catalog No. 0218398801 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mL
10 mL
100 mL
1000 mL

Ready-to-use proteinase K solution

  • Fungus-Derived Proteinase K Purified Recombinant
  • Prepared at 20 mg/mL in 10 mM Tris-HCl, 1 mM calcium acetate, pH 7.5 containing 50% glycerol
  • Molecular Biology Grade
  • Extremely effective on native proteins
  • Can be used to rapidly inactivate endogenous nucleases such as RNases and DNases
  • Sterile filtered using 0.22 μm filter
  • Can maintain its activity for a long time at room temperature and low temperature
  • Store at 2-8°C for long-term storage
TRUSTED_SUSTAINABILITY

Specifications

Characterization Mixtures
Concentration 20 mg/mL
For Use With (Application) Nuclease Inactivation
Formulation 20 mg/mL in 10 mM Tris-HCl, 1 mM calcium acetate, pH 7.5 containing 50% glycerol
Molecular Weight (g/mol) 29 kDa
Preparation Method Working concentrations of the enzyme range from 50 to 250 μg/mL.
Quantity 1 mL
Storage Requirements 2 to 8°C
Water Content 0.478
Endotoxin Concentration 29 kDa
Recombinant Recombinant
Classification GHS08
Specific Activity >40 U/mg protein
Technique PCR, RNA Purification, DNA Purification, NGS, Sample Preparation
Expression System Fungus
Form Liquid
Color Yellowish-green
Purity or Quality Grade >95%
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.