Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ProteoExtract™ Complete Mammalian Proteome Extraction Kit

Catalog No. 5397791kit Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 kit

Designed to extract virtually all proteins from both mammalian tissues and culture cells

  • A simple two step process within a microcentrifuge tube to minimize protein loss
  • The extraction results in a total protein sample that is a clarified and non-viscous protein solution showing higher resolution and an increased total number of spots in 2-D gel electrophoresis
  • Sample size: 1x107 - 1x108 cells or 50-100mg tissue
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kit
Format 20 Extractions
Kit Contents Wash buffer, Resuspension buffer, Extraction reagent, Reducing reagent, Benzonase nuclease, Glass beads, and user protocol
Content And Storage Store at+2°C to+8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.