Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Proteome Profiler Human NFkB Pathway Array



Click to view available options
Quantity:
1 Kit
Description
The Proteome Profiler Human NFkB Pathway Array contains 4 membranes-each spotted in duplicate with 41 different proteins and 4 serine or tyrosine phosphorylation sites antibodies.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Description | NFkB Pathway Array Kit |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Quantity | 1 Kit |
For Use With (Application) | For parallel determination of human NFkB Pathway proteins |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction