Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Proteome Profiler Human Phospho-Kinase Array Kit
GREENER_CHOICE

Catalog No. ary003c
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Contains 4 sets of 2 membranes - each spotted in duplicate with antibodies against 37 different kinases and 2 related total proteins

Analyzing the phosphorylation profiles of kinases and their protein substrates is essential for understanding how cells recognize and respond to changes in their environment. The Human Phospho-Kinase Array is a rapid, sensitive, and economical tool to simultaneously detect the relative levels of phosphorylation of 37 kinase phosphorylation sites and 2 related total proteins without performing numerous immunoprecipitations and Western blots. Each capture antibody was carefully selected using cell lysates prepared from cell lines known to express the target protein.

Specifications

Content And Storage Store the unopened product at 2 - 8°C. Do not use past expiration date.
Quantity 1 Kit
Target Species Human
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.