Learn More
R&D Systems™ Proteome Profiler Human Protease/Protease Inhibitor Array
GREENER_CHOICE
Catalog No. ary025
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
Catalog No. ARY025
Supplier: R&D Systems™ ARY025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Proteome Profiler Human Protease/Protease Inhibitor Array requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Protease/Protease Inhibitor Array contains 4 membranes-2 spotted in duplicate with antibodies against 35 different proteases and 2 spotted in duplicate with antibodies against 32 different protease inhibitors.
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Kit Contents 4 Array Membranes (2 of Proteases and 2 Protease Inhibitors), 4-Well Multi-Dish, Array Buffers, Wash Buffer, Detection Antibody Cocktail, Human Protease Array, Detection Antibody Cocktail, Human Protease Inhibitor Array, Streptavidin-HRP, Chemiluminescent Detection Reagents, Transparency Overlay Template, Detailed Protocol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.