Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prothrombin Fragment 1/2 Monoclonal Antibody (D3), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA550945
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550945 200 μL
FEMA550946 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550945 Supplier Invitrogen™ Supplier No. MA550945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prothrombin Fragment 1/2 is a protein fragment that is produced by the cleavage of prothrombin, a blood clotting protein. It is a small peptide consisting of the first half of the prothrombin molecule and is involved in the formation of blood clots. Prothrombin Fragment 1/2 is used as a biomarker for the diagnosis and monitoring of various blood clotting disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prothrombin Fragment 1/2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene F2
Gene Accession No. P00734, Q19AZ8
Gene Alias prothrombin fragment 1.2; Prothrombin fragment 1+2; prothrombin fragment F1.2; Prothrombin fragment F1+2
Gene Symbols F2
Host Species Mouse
Immunogen Recombinant Human F1+2 protein, Ala44-Arg327 (Accession #P00734).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100144442, 2147
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.