Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prothrombin Monoclonal Antibody (C13)
GREENER_CHOICE

Catalog No. PIMA541567
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541567 200 μL
PIMA543933 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541567 Supplier Invitrogen™ Supplier No. MA541567
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Prothrombin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prothrombin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene F2
Gene Accession No. P18292
Gene Alias Activation peptide fragment 1; Activation peptide fragment 2; bovine prothrombin protein; Cf2; Cf-2; coagulation factor 2; coagulation factor II; coagulation factor II (thrombin); coagulation factor II [thrombin]; coagulation factor II, thrombin; F2; FII; kringle-2 domain; fragment-2; prepro-coagulation factor II; preprothrombin; prothrombin; prothrombin B-chain; prothrombin precursor; PT; RPRGL2; serine protease; THPH1; THROMBIN; Thrombin heavy chain; Thrombin light chain
Gene Symbols F2
Host Species Mouse
Immunogen Recombinant protein Coagulation Factor II. The antigen corresponds to amino acid range 201-323 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29251
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.