Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prothrombin Monoclonal Antibody (C19), Invitrogen™

Catalog No. FEMA550986
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550986 20 μL
FEMA550985 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550986 Supplier Invitrogen™ Supplier No. MA550986
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prothrombin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C19
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene F2
Gene Accession No. P18292
Gene Alias Activation peptide fragment 1; Activation peptide fragment 2; bovine prothrombin protein; Cf2; Cf-2; coagulation factor 2; coagulation factor II; coagulation factor II (thrombin); coagulation factor II [thrombin]; coagulation factor II, thrombin; F2; FII; kringle-2 domain; fragment-2; prepro-coagulation factor II; preprothrombin; prothrombin; prothrombin B-chain; prothrombin precursor; PT; RPRGL2; serine protease; THPH1; THROMBIN; Thrombin heavy chain; Thrombin light chain
Gene Symbols F2
Host Species Mouse
Immunogen Recombinant Rat F2 protein, Ser201-Arg323 (Accession #P18292).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29251
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.