Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Prothrombin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579226
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human plasma. IHC: human colon cancer tissue, human lung cancer tissue. Flow: Human PBMC cell.
Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.
Specifications
Prothrombin | |
Polyclonal | |
Unconjugated | |
F2 | |
Activation peptide fragment 1; Activation peptide fragment 2; bovine prothrombin protein; Cf2; Cf-2; coagulation factor 2; coagulation factor II; coagulation factor II (thrombin); coagulation factor II [thrombin]; coagulation factor II, thrombin; F2; FII; kringle-2 domain; fragment-2; prepro-coagulation factor II; preprothrombin; prothrombin; prothrombin B-chain; prothrombin precursor; PT; RPRGL2; serine protease; THPH1; THROMBIN; Thrombin heavy chain; Thrombin light chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2147 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P00734 | |
F2 | |
E. coli-derived human Prothrombin recombinant protein (Position: Y67-R124). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction