Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protocadherin-1 Antibody (648127), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5899Y
Description
Protocadherin-1 Monoclonal antibody specifically detects Protocadherin-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
Protocadherin-1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5097 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, CyTOF | |
648127 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
cadherin-like 1, Cadherin-like protein 1, FLJ53887, MGC45991, pc42, PC42PCDH42, protocadherin 1, protocadherin 1 (cadherin-like 1), protocadherin 42, protocadherin-1, protocadherin-42 | |
E. coli-derived recombinant human Protocadherin-1, Thr58-Asn162, Accession # Q08174 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction