Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protocadherin-8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Protocadherin-8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Protocadherin-8 Polyclonal specifically detects Protocadherin-8 in Human samples. It is validated for Western Blot.Specifications
Protocadherin-8 | |
Polyclonal | |
Rabbit | |
ARCADLIN, PAPC, protocadherin 8, protocadherin-8 | |
PCDH8 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
5100 | |
Synthetic peptides corresponding to PCDH8(protocadherin 8) The peptide sequence was selected from the middle region of PCDH8. Peptide sequence RVFRALLVISDGGRPPLTTTATVSFVVTAGGGRGPAAPASAGSPERSRPP. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title