Learn More
Protocadherin gamma C3 Antibody (926518), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB83641MFV610
Description
Protocadherin gamma C3 Monoclonal antibody specifically detects Protocadherin gamma C3 in Human samples. It is validated for Flow CytometrySpecifications
| Protocadherin gamma C3 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
| 5098 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| 926518 | |
| Flow Cytometry | |
| cadherin-like 2, PC43, PC-43, PCDH2, PCDH-GAMMA-C3, protocadherin 2, protocadherin 43, protocadherin gamma subfamily C, 3, protocadherin gamma-C3, protocadherin-2, Protocadherin-43 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin gamma C3, Met1-Tyr693, Accession # Q9UN70 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |