Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protocadherin gamma C3 Antibody (926518), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB83641PEATT594
Description
Protocadherin gamma C3 Monoclonal antibody specifically detects Protocadherin gamma C3 in Human samples. It is validated for Flow CytometrySpecifications
Protocadherin gamma C3 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
926518 | |
Flow Cytometry | |
cadherin-like 2, PC43, PC-43, PCDH2, PCDH-GAMMA-C3, protocadherin 2, protocadherin 43, protocadherin gamma subfamily C, 3, protocadherin gamma-C3, protocadherin-2, Protocadherin-43 | |
Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin gamma C3, Met1-Tyr693, Accession # Q9UN70 | |
0.1 mL | |
Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
5098 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction