Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protocadherin Gamma Monoclonal Antibody (N159/5), PE
GREENER_CHOICE

Catalog No. PIMA545661
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545661 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545661 Supplier Invitrogen™ Supplier No. MA545661
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45661 was sufficient for detection of Protocadherin gamma (pan) in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 100kDa. Cross-reacts with all Gamma-protocadherin-A, -B and -C proteins. This antibody was formerly sold as clone S159-5.

The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protocadherin Gamma
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N159/5
Concentration 1 mg/mL
Conjugate PE
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene PCDHGA12
Gene Accession No. O60330, Q91XY4, Q9UN70, Q9UN71, Q9Y5F6, Q9Y5F7, Q9Y5F8, Q9Y5F9, Q9Y5G0, Q9Y5G1, Q9Y5G2, Q9Y5G3, Q9Y5G4, Q9Y5G5, Q9Y5G6, Q9Y5G7, Q9Y5G8, Q9Y5G9, Q9Y5H0, Q9Y5H1, Q9Y5H2, Q9Y5H3, Q9Y5H4
Gene Alias 3' RACE product; cadherin 20; cadherin 21; cadherin ME3; cadherin ME6; cadherin-20; Cadherin-21; cadherin-like 2; CDH20; CDH21; CDH-GAMMA-A5; FIB2; FIB3; fibroblast cadherin FIB2; fibroblast cadherin FIB3; fibroblast cadherin-2; Fibroblast cadherin-3; KIAA0327; KIAA0588; ME3; ME6; mKIAA0588; mKIAA4054; Pan-Gamma-Protocadherin-A; pc2c; PC43; PC-43; PCDH gamma A; Pcdh13; PCDH2; PCDHGA; PCDHGA1; PCDHGA10; PCDHGA11; PCDHGA12; Pcdhga2; PCDHGA3; Pcdhga4; PCDHGA5; PCDHGA6; PCDHGA7; Pcdhga8; PCDHGA9; PCDH-gamma-A; PCDH-GAMMA-A1; PCDH-gamma-A10; PCDH-GAMMA-A11; PCDH-gamma-A12; PCDH-GAMMA-A2; PCDH-gamma-A3; PCDH-gamma-A4; PCDH-GAMMA-A5; PCDH-gamma-A6; PCDH-GAMMA-A7; PCDH-gamma-A8; PCDH-gamma-A9; PCDH-GAMMA-B1; PCDH-GAMMA-B2; PCDH-gamma-B3; PCDH-GAMMA-B4; PCDH-GAMMA-B5; PCDH-GAMMA-B6; PCDH-gamma-B7; PCDH-GAMMA-C3; PCDH-gamma-C4; PCDH-gamma-C5; Pcdhgb1; PCDHGB2; PCDHGB3; PCDHGB4; PCDHGB5; Pcdhgb6; Pcdhgb7; PCDHGC3; Pcdhgc4; PCDHGC5; protocadherin 13; protocadherin 2; protocadherin 43; protocadherin gamma A12; protocadherin gamma B2; protocadherin gamma B6; protocadherin gamma C-VI; protocadherin gamma subfamily A, 1; protocadherin gamma subfamily A, 10; protocadherin gamma subfamily A, 11; protocadherin gamma subfamily A, 12; protocadherin gamma subfamily A, 2; protocadherin gamma subfamily A, 3; protocadherin gamma subfamily A, 4; protocadherin gamma subfamily A, 5; protocadherin gamma subfamily A, 6; protocadherin gamma subfamily A, 7; protocadherin gamma subfamily A, 8; protocadherin gamma subfamily A, 9; protocadherin gamma subfamily B, 1; protocadherin gamma subfamily B, 1, isoform 2; protocadherin gamma subfamily B, 2; protocadherin gamma subfamily B, 3; protocadherin gamma subfamily B, 4; protocadherin gamma subfamily B, 5; protocadherin gamma subfamily B, 6; protocadherin gamma subfamily B, 7; protocadherin gamma subfamily C, 3; protocadherin gamma subfamily C, 4; protocadherin gamma subfamily C, 5; protocadherin gamma-A1; Protocadherin gamma-A10; protocadherin gamma-A11; Protocadherin gamma-A12; protocadherin gamma-A2; Protocadherin gamma-A3; protocadherin gamma-A4; Protocadherin gamma-A5; protocadherin gamma-A6; Protocadherin gamma-A7; protocadherin gamma-A8; Protocadherin gamma-A9; protocadherin gamma-B1; Protocadherin gamma-B2; protocadherin gamma-B3; Protocadherin gamma-B4; protocadherin gamma-B5; Protocadherin gamma-B6; protocadherin gamma-B7; Protocadherin gamma-C3; protocadherin gamma-C4; Protocadherin gamma-C5; protocadherin-2; Protocadherin-43; UNQ371/PRO707
Gene Symbols Pcdhga1, Pcdhga10, Pcdhga11, PCDHGA12, Pcdhga2, PCDHGA3, PCDHGA4, Pcdhga5, PCDHGA6, Pcdhga7, PCDHGA8, Pcdhga9, PCDHGB1, PCDHGB2, PCDHGB3, Pcdhgb4, Pcdhgb5, Pcdhgb6, Pcdhgb7, Pcdhgc3, PCDHGC4, PCDHGC5
Host Species Mouse
Immunogen Fusion protein corresponding to amino acids 808-931 (C-terminal cytoplasmic constant domain) of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins (A subfamily amino acids ~807-930, B subfamily amino acids ~789-912 and C subfamily amino acids ~818-941).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102552497, 102552705, 102552796, 116782, 252894, 252895, 252897, 26025, 291635, 291637, 364843, 364844, 498846, 498847, 498849, 498850, 5098, 553129, 56097, 56098, 56099, 56100, 56101, 56102, 56103, 56104, 56105, 56106, 56107, 56108, 56109, 56110, 56111, 56112, 56113, 56114, 641525, 641526, 680361, 680448, 8641, 93699, 93700, 93701, 93702, 93703, 93704, 93706, 93707, 93708, 93709, 93710, 93711, 93712, 93713, 93714, 93715, 93716, 93717, 93722, 93723, 93724, 9708
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.