Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protocadherin gamma (pan) Monoclonal Antibody (S159-5), QED Bioscience™
SDP

Catalog No. 502278241
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8241 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8241 Supplier Leinco Technologies Inc Supplier No. 56562100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Protocadherin gamma (pan) Monoclonal antibody specifically detects Protocadherin gamma (pan) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen Protocadherin gamma (pan)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone S159-5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Gene PCDHGA12
Gene Accession No. O60330, Q91XY4, Q9UN70, Q9UN71, Q9Y5F6, Q9Y5F7, Q9Y5F8, Q9Y5F9, Q9Y5G0, Q9Y5G1, Q9Y5G2, Q9Y5G3, Q9Y5G4, Q9Y5G5, Q9Y5G6, Q9Y5G7, Q9Y5G8, Q9Y5G9, Q9Y5H0, Q9Y5H1, Q9Y5H2, Q9Y5H3, Q9Y5H4
Gene Alias 3' RACE product; cadherin 20; cadherin 21; cadherin ME3; cadherin ME6; cadherin-20; Cadherin-21; cadherin-like 2; CDH20; CDH21; CDH-GAMMA-A5; FIB2; FIB3; fibroblast cadherin FIB2; fibroblast cadherin FIB3; fibroblast cadherin-2; Fibroblast cadherin-3; KIAA0327; KIAA0588; ME3; ME6; mKIAA0588; mKIAA4054; Pan-Gamma-Protocadherin-A; pc2c; PC43; PC-43; PCDH gamma A; Pcdh13; PCDH2; PCDHGA; PCDHGA1; PCDHGA10; PCDHGA11; PCDHGA12; Pcdhga2; PCDHGA3; Pcdhga4; PCDHGA5; PCDHGA6; PCDHGA7; Pcdhga8; PCDHGA9; PCDH-gamma-A; PCDH-GAMMA-A1; PCDH-gamma-A10; PCDH-GAMMA-A11; PCDH-gamma-A12; PCDH-GAMMA-A2; PCDH-gamma-A3; PCDH-gamma-A4; PCDH-GAMMA-A5; PCDH-gamma-A6; PCDH-GAMMA-A7; PCDH-gamma-A8; PCDH-gamma-A9; PCDH-GAMMA-B1; PCDH-GAMMA-B2; PCDH-gamma-B3; PCDH-GAMMA-B4; PCDH-GAMMA-B5; PCDH-GAMMA-B6; PCDH-gamma-B7; PCDH-GAMMA-C3; PCDH-gamma-C4; PCDH-gamma-C5; Pcdhgb1; PCDHGB2; PCDHGB3; PCDHGB4; PCDHGB5; Pcdhgb6; Pcdhgb7; PCDHGC3; Pcdhgc4; PCDHGC5; protocadherin 13; protocadherin 2; protocadherin 43; protocadherin gamma A12; protocadherin gamma B2; protocadherin gamma B6; protocadherin gamma C-VI; protocadherin gamma subfamily A, 1; protocadherin gamma subfamily A, 10; protocadherin gamma subfamily A, 11; protocadherin gamma subfamily A, 12; protocadherin gamma subfamily A, 2; protocadherin gamma subfamily A, 3; protocadherin gamma subfamily A, 4; protocadherin gamma subfamily A, 5; protocadherin gamma subfamily A, 6; protocadherin gamma subfamily A, 7; protocadherin gamma subfamily A, 8; protocadherin gamma subfamily A, 9; protocadherin gamma subfamily B, 1; protocadherin gamma subfamily B, 1, isoform 2; protocadherin gamma subfamily B, 2; protocadherin gamma subfamily B, 3; protocadherin gamma subfamily B, 4; protocadherin gamma subfamily B, 5; protocadherin gamma subfamily B, 6; protocadherin gamma subfamily B, 7; protocadherin gamma subfamily C, 3; protocadherin gamma subfamily C, 4; protocadherin gamma subfamily C, 5; protocadherin gamma-A1; Protocadherin gamma-A10; protocadherin gamma-A11; Protocadherin gamma-A12; protocadherin gamma-A2; Protocadherin gamma-A3; protocadherin gamma-A4; Protocadherin gamma-A5; protocadherin gamma-A6; Protocadherin gamma-A7; protocadherin gamma-A8; Protocadherin gamma-A9; protocadherin gamma-B1; Protocadherin gamma-B2; protocadherin gamma-B3; Protocadherin gamma-B4; protocadherin gamma-B5; Protocadherin gamma-B6; protocadherin gamma-B7; Protocadherin gamma-C3; protocadherin gamma-C4; Protocadherin gamma-C5; protocadherin-2; Protocadherin-43; UNQ371/PRO707
Host Species Mouse
Immunogen Fusion protein corresponding to aa 808-931 (C-terminal cytoplasmic constant domain) of mouse PCDHGA1. This sequence is present in all 22 PCDHG proteins.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102552497, 102552705, 102552796, 116782, 252894, 252895, 252897, 26025, 291635, 291637, 364843, 364844, 498846, 498847, 498849, 498850, 5098, 553129, 56097, 56098, 56099, 56100, 56101, 56102, 56103, 56104, 56105, 56106, 56107, 56108, 56109, 56110, 56111, 56112, 56113, 56114, 641525, 641526, 680361, 680448, 8641, 93699, 93700, 93701, 93702, 93703, 93704, 93706, 93707, 93708, 93709, 93710, 93711, 93712, 93713, 93714, 93715, 93716, 93717, 93722, 93723, 93724, 9708
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.