Learn More
Protocol™ Decalcifying Solution B
Click to view available options
Quantity:
32 oz.
1 gal.
Description
- Can be used on all types of calcified tissue and is especially effective on bone marrow samples
- Cellular and nuclear details remain crisp and clear
Specifications
Specifications
Product Type | Decalcifying Solution |
Quantity | 1 gal. |
Solution Type | Decalcifier |
Form | Solution |
Made in the USA