Learn More
Fisher Healthcare™ PROTOCOL™ Parasitology Three-Vial Kits, SAF/Clean/C and S
Shop All Fisherbrand ProductsDescription
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Three-Vial Kits.
Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA)
- Mercury-free fixative for use as a preservative/fixative for protozoan trophozoites
- Slides fixed with Zn-PVA may be stained using trichrome or hematoxylin
Sodium Acetate Acetic Acid Formalin (SAF)
- Mercury-free preservative/fixative ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
- May be stained using trichrome or hematoxylin
10% Buffered Formalin
- All-purpose fixative appropriate for helminth eggs and larvae, and for protozoan cysts
- Buffering system helps to maintain organism morphology
- Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts
Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA)
- Acts as a preservative/fixative for protozoan trophozoites
- Slides fixed with Modified PVA may be stained using trichrome or hematoxylin
Culture and Sensitivity (C & S Media)
- For collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter, and CholeraVibrio
- Bacterial population is maintained for optimum recovery, up to 96 hours
Clean Vials
- Empty, 30mL vials for transporting specimens
Ordering Alerts:
Supplied as 10 packs of three vials each
Note:
All PROTOCOL Parasitology products are mercury free.
Specifications
Specifications
| Product Type | Parasitology Three Vial Kit |
| Media Type | Zn-PVA, Formalin, C&S |
| Quantity | 10 Kits/Pk. |
| Volume (English) | 1.01 oz. |
| Volume (Metric) | 30 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.