Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protogenin Antibody (OTI2B3), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273674AF594
Description
Protogenin Monoclonal specifically detects Protogenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Protogenin | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PRTG | |
Full length recombinant protein of human Protogenin (NP_776175) produced in HEK293T cells. | |
0.1 mL | |
283659.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2B3 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
FLJ12795, FLJ13221, FLJ25756, IGDCC5, immunoglobulin superfamily, DCC subclass, member 5, Protein Shen-Dan, protogenin, protogenin homolog (Gallus gallus), shen-dan | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction