Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protogenin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547671
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547671 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547671 Supplier Invitrogen™ Supplier No. PA547671
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Protogenin, encoded by the PRTG gene, is a cell surface protein that belongs to the immunoglobulin superfamily and plays a critical role in neural development. Protogenin is involved in cell adhesion and signaling pathways that regulate processes such as neurogenesis, neuronal migration, and the establishment of neural circuits during embryonic development. Its expression is predominantly observed in the developing nervous system, where it facilitates interactions between cells and the extracellular matrix, contributing to the proper formation of brain structures. Protogenin's function in regulating cellular communication is essential for maintaining the balance between stem cell renewal and differentiation in neural tissues. Dysregulation or mutations in the PRTG gene can lead to developmental abnormalities and have been investigated for their potential connections to neurodevelopmental disorders. Research continues to explore protogenin's mechanisms and interaction networks to better understand its contributions to neural development and its potential as a therapeutic target in developmental disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protogenin
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PRTG
Gene Accession No. Q2EY15
Gene Alias A230098A12Rik; IGDCC5; immunoglobulin superfamily, DCC subclass, member 5; protein Shen-Dan; Protogenin; protogenin homolog (Gallus gallus); prtg; shen-dan
Gene Symbols PRTG
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PRTG Val24-Thr943.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235472
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.