Learn More
StatLab Protozoa Suspension
SDP
Catalog No. p-200110338
Click to view available options
Species:
Escherichia coli
Giardia lamblia
Iodamoeba
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Excellent laboratory parasitology control material.
  • Utilizes simulated clinical control material with formalin-fixed suspensions of common protozoa
  • All of our rare teaching materials are subject to availability
  • 1 cc vial of protozoa suspension, choose between E. coli, Giardia Lamblia, and Iodamoeba
Specifications
Product Type Protozoa Suspension
Sterility Non-sterile
Species Escherichia coli
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.