Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prox1 Antibody (5G10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP130045W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130045W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130045W Supplier Novus Biologicals Supplier No. NBP130045AF405
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prox1 Monoclonal specifically detects Prox1 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount.

Specifications

Antigen Prox1
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5G10
Conjugate Alexa Fluor 405
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q92786
Gene Alias Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1
Gene Symbols PROX1
Host Species Mouse
Immunogen The highly conserved Homeo and Prospero domain of human Prox1. [UniProt# Q92786]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 5629
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.