Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prox1 Antibody (5G10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280920
Description
Prox1 Monoclonal specifically detects Prox1 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount, CyTOF-ready.Specifications
Prox1 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry, ELISA, Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence : 1:200, Immunohistochemistry-Paraffin : 1:100-1:1000, Immunohistochemistry Whole-Mount reported in scientific literature (PMID 24654984), CyTOF-ready | |
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
It is expected to work with all vertebrates including mammals, birds and fish. Please note that this antibody is reactive to Mouse and derived from the same host, Mouse. Additional Mouse on Mouse blocking steps may be required for IHC and ICC experiments. Please contact Technical Support for more information. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
5G10 | |
Unconjugated | |
PBS | |
PROX1 | |
The highly conserved Homeo and Prospero domain of human Prox1. [UniProt# Q92786] | |
0.1 mL | |
Vision | |
5629 | |
Human, Mouse, Rat, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction