Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prox1 Antibody (5G10), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130045MFV610
Description
Prox1 Monoclonal antibody specifically detects Prox1 in Human, Mouse, Rat, Chicken samples. It is validated for Flow CytometrySpecifications
Prox1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Flow Cytometry | |
5G10 | |
Flow Cytometry | |
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1 | |
The highly conserved Homeo and Prospero domain of human Prox1. [UniProt# Q92786] | |
0.1 mL | |
Vision | |
5629 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction