Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prox1 Antibody (CL13474), Novus Biologicals™

Mouse Monoclonal Antibody
$342.50 - $534.50
Specifications
Antigen | Prox1 |
---|---|
Clone | CL13474 |
Dilution | Immunocytochemistry/ Immunofluorescence 2-10 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Description
Prox1 Monoclonal antibody specifically detects Prox1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Prox1 | |
Immunocytochemistry/ Immunofluorescence 2-10 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1 | |
This antibody was designed to target the recombinant protein Prox1, with the precise immunogen sequence being withheld for proprietary reasons. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL13474 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Vision | |
PBS, pH 7.2, 40% glycerol | |
5629 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title