Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROX1 Mouse anti-Human, Rat, Clone: 5G10, Abnova™

Catalog No. 89035543 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-543 Supplier Abnova Corporation Supplier No. MAB14145

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against recombinant human PROX1.

  • High specificity
  • High reactivity

Specifications

Antigen PROX1
Applications Immunohistochemistry
Classification Monoclonal
Clone 5G10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human PROX1.
Dilution Immunohistochemistry The optimal working dilution should be determined by the end user.
Formulation In HEPES, 0.15 M NaCl, pH 7.5 (50% glycerol, 100 ug/mL BSA).
Gene Accession No. Q92786
Gene Symbols PROX1
Host Species Mouse
Immunogen Recombinant protein corresponding to human PROX1.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5629
Target Species Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.