Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PROX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560913 Supplier Invitrogen™ Supplier No. PA560913
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLLDPPGHLT QLGRSFQGQV AEGRSEPSPP VGGACKDPLA LAALPRRVQL QAGVPVGNLS LAKRLDSPRY PIPPRMTPKP CQDP Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 58%.

Transcription regulator. Does not seem to be essential for embryonic development and postnatal survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROX2
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PROX2
Gene Accession No. Q3B8N5
Gene Alias 1700058C01Rik; Homeobox prospero-like protein PROX2; prospero homeobox 2; prospero homeobox protein 2; PROX2; PROX-2; RGD1310315
Gene Symbols PROX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PROX2. Recombinant protein control fragment (Product #RP-100454).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283571
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.