Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Proximal Nephrogenic Renal Antigen Monoclonal Antibody (PN-15 (66.4.C2))
GREENER_CHOICE

Catalog No. MA135774
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA135774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA135774 Supplier Invitrogen™ Supplier No. MA135774
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PN-15 reacts with a lectin receptor like glycoprotein of 200 kDa (gp200), present in proximal renal tubules and on urothelium. The antigen is carbohydrate in nature. Other normal tissues that display the antigen include breast, parathyroid glands, thymus and epididymis. Among renal carcinomas 93% of primary and 84% of metastatic carcinomas are positive. Bladder cancers are also largely positive. Other tumor types include breast cancer, teratocarcinomas and parathyroid adenomas. The antigen, also called DEC-205, was assigned to CD205 at CD workshop VII. In the immune system it can facilitate tolerance to self-antigens through uptake of apoptosis derived material by dendritic cells, which in turn present fragments through MHC II and MHC I, either inducing or repressing immune responses, depending on the nature of concomitant signals.

PNRA is a 200 kDa glycoprotein found in proximal renal tubules, and may be useful in characterizing renal cell carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proximal Nephrogenic Renal Antigen
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone PN-15 (66.4.C2)
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias PNRA
Host Species Mouse
Immunogen Renal cortical tissue extract.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rat, Monkey, Horse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.