Learn More
Honeywell™ Proximity Firefighting Gloves - Gauntlet Cuff
Catalog No. p-2450308
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Knit PBI and Aramid Fiber based aluminized material is the preferred shell for proximity turnout gear due to its flexibility, durability, and radiant heat reflectivity.
  • Knit Pbi and Aramid Fiber based aluminized material is the preferred shell for Proximity turnout gear
  • Elkskin leather palm increases heat protection when gripping hot objects
  • Crosstech moisture barrier helps keep hands dry and comfortable
  • Aramid Fiber/Nomex™ lining for increased burn resistance
  • Supplemental wool felt insulation inside of the glove provides an additional layer of protection
Certifications
NFPA 1971-2007
TRUSTED_SUSTAINABILITY
Specifications
Back Material Aluminized PBI/Aramid Fiber
Certifications/Compliance NFPA 1971-2007
Coating Material Uncoated
Color Brown, Silver
Hand Specific
Product Type Proximity Firefighting Gloves
Size Small
Glove Cuff Color Silver
Glove Cuff Style Gauntlet
Liner Material Nomex™/Aramid Fiber
Palm Type Elkhide Leather
Series GL-BPR-RGA
Thumb Style Wing
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.