Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PrP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1750 Supplier Invitrogen™ Supplier No. PA1750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-750 detects prion protein (PrP) from sheep tissues. PA1-750 has been successfully used in Western blot, immunoprecipitation and immunohistochemistry procedures. By Western blot, this antibody detects a 33-35 kDa protein representing PrPC and a 27-30 kDa protein representing PrPSc from sheep normal and infected brains. Immunohistochemical staining of PrP in formalin-fixed infected sheep brain with PA1-750 results in intense staining of PrP plaques which are known to exhibit an abnormally high concentration of PrP. PA1-750 immunizing peptide corresponds to amino acid residues 90-102 from hamster PrP. PA1-750 immunizing peptide (Cat. # PEP-034) is available for use in neutralization and control experiments.

Human prion protein (PrP), also known as PRNP, is a ubiquitously expressed GPI-anchored cell surface glycoprotein associating with lipid raft components and functioning as a signaling molecule. PrP plays a role in apoptosis in a cell context-dependent manner, is involved in proliferation of epithelial cells and in distribution of junction-associated proteins in human enterocytes. Conversion of this normal cellular prion protein (PrPc) into an abnormal conformer (PrPSc) is the crucial step associated with triggering the pathogenesis of the prion neurodegenerative disorders, such as the Creutzfeld-Jakob disease (CJD). Whereas PrPc is rich in alpha-helices, the PrPSc form has higher content of beta-sheets and is resistant to proteinase K.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PrP
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene PRNP
Gene Accession No. P23907
Gene Alias AA960666; AI325101; Alternative prion protein; alternative prion protein; major prion protein; AltPrP; ASCR; BSE; CD230; CD230 antigen; CJD; GSS; KURU; major prion protein; major prion protein; alternative prion protein; major scrapie-associated fibril protein 1; MGC26679; p27-30; PK resistant core; prion protein; prion protein (p27-30) (Creutzfeldt-Jakob disease, Gerstmann-Strausler-Scheinker syndrome, fatal familial insomnia); prion protein precursor; prion protein precursor PrP; prion protein PrP; prion protein variant a; prion protein variant b; prion protein, PrP; prion protein, structural; prion-related protein; PRIP; Prn; Prn-i; Prnp; Prn-p; PrP; PrP>C>; PrP27-30; PrP33-35C; PrPc; PrPSc; Sinc; SIP; TSE
Gene Symbols PRNP
Host Species Rabbit
Immunogen Synthetic Peptide: G(90) Q G G G T H N Q W N K P(102) G G C.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493887
Target Species Sheep
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.