Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRP19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529254
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529254 Supplier Invitrogen™ Supplier No. PA529254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Molt-4. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (83%), Pig (100%), Chicken (97%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PSO4 is the human homolog of yeast Pso4, a gene essential for cell survival and DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRP19
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Prpf19
Gene Accession No. Q99KP6, Q9UMS4
Gene Alias AA617263; AL024362; D19Wsu55e; hPSO4; Neuronal differentiation-related gene protein; NMP200; Nuclear matrix protein 200; nuclear matrix protein NMP200 related to splicing factor PRP19; nuclear matrix protein SNEV; pre-mRNA processing factor 19; pre-mRNA-processing factor 19; PRP19; prp19 beta; PRP19/PSO4 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae); PRPF19; PSO4; psoralen 4; RING-type E3 ubiquitin transferase PRP19; Senescence evasion factor; SNEV; UBOX4
Gene Symbols Prpf19
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRP19. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27339, 28000
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.