Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRP19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519550
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519550 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519550 Supplier Thermo Scientific Supplier No. PA519550
Add to Cart
Add to Cart

PRP19 Polyclonal Antibody

PRP19 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PRP19
Applications Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene Prpf19
Gene Accession No. Q99KP6, Q9JMJ4, Q9UMS4
Gene Alias AA617263; AL024362; D19Wsu55e; hPSO4; Neuronal differentiation-related gene protein; NMP200; Nuclear matrix protein 200; nuclear matrix protein NMP200 related to splicing factor PRP19; nuclear matrix protein SNEV; pre-mRNA processing factor 19; pre-mRNA-processing factor 19; PRP19; prp19 beta; PRP19/PSO4 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae); PRPF19; PSO4; psoralen 4; RING-type E3 ubiquitin transferase PRP19; Senescence evasion factor; SNEV; UBOX4
Gene Symbols Prpf19
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 450 to the C-terminus of Human PRP19.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246216, 27339, 28000
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.