Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561428 Supplier Invitrogen™ Supplier No. PA561428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARNTLMDMRL SQVSDSVSGQ TVVDPKGYLT DLNSMIPTHG GDINDIKKAR LLLKSVRETN PHHPPAWIAS ARLEEVTGKL QVARNL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRP6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRPF6
Gene Accession No. O94906
Gene Alias 1190003A07Rik; 2610031L17Rik; androgen receptor N-terminal domain transactivating protein-1; androgen receptor N-terminal domain-transactivating protein 1; ANT1; ANT-1; C20orf14; hPrp6; p102 U5 small nuclear ribonucleoprotein particle-binding protein; pre-mRNA processing factor 6; pre-mRNA-processing factor 6; Prp6; PRP6 homolog; PRP6 pre-mRNA processing factor 6 homolog; PRP6 pre-mRNA splicing factor 6 homolog; PRP6 pre-mRNA splicing factor 6 homolog (yeast); Prpf6; putative mitochondrial outer membrane protein import receptor; RGD1307103; RP60; SNRNP102; TOM; U5 snRNP-associated 102 kDa protein; U5-102 kDa protein; U5-102K
Gene Symbols PRPF6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRP6. Recombinant protein control fragment (Product #RP-99302).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24148
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.